Learn More
Applied Biosystems™ MicroSEQ™ ID Fungal Gene Library v2022
Product Code. 18733634 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity unitSize
18733634 1 Ea. Each
1 options
Product Code. 18733634 Supplier Applied Biosystems™ Supplier No. A57314
The MicroSEQ ID Fungal Gene Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0.
The MicroSEQ ID Fungal Gene Library v2022 is for use with MicroSEQ ID Microbial Identification Software v4.0. It contains over 1300 reference sequences from the D2 region of the Large Subunit (rDNA) of yeast and mold species against which an unknown yeast or mold sequence can be compared in order to identify the unknown organism.
TRUSTED_SUSTAINABILITY
Specifications
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) SeqStudio Genetic Analyzers, SeqStudio Flex Genetic Analyzers
For Use With (Application) Microbial Identification
License Static
Operating System Microsoft Windows 10
Product Line MicroSeq™ ID
Product Type Microbial Identification Software
Quantity 1 Ea.
Software Data Collection and Analysis Software