missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit

Product Code. 18705525 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
96 samples
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18705525 96 samples Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18705525 Supplier Applied Biosystems™ Supplier No. A33246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit is used for post-sequencing clean-up in the MicroSEQ ID workflow.

The MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit is used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel-filtration matrix used in the strip format provides a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions.

The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit can be used downstream of the following sequencing kits:

  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit

The kit consists of twelve 8-well strips and a 96-well receiver plate to accommodate 96 sequencing samples. It is ideal for low- to medium-throughput microbial-identification workflows. The end products can be used directly for sequencing on any Applied Biosystems™ genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 12 x 8-strip clean-up tubes
• 2 x 96-well waste plate
• 2 x 96-well receiver plate
• 1 x 8-strip balancing strip

Format Strip
Sample Type Sequencing Products
For Use With (Application) Sequencing
High-throughput Compatibility Manual Protocols
No. of Samples 96 Samples
Product Line MicroSEQ™ ID
Quantity 96 samples
Target Fluorescent Dye Removal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.