missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Refill Kit

Product Code. 18725425 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
96 Samples
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18725425 96 Samples Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18725425 Supplier Applied Biosystems™ Supplier No. A33256
  • ONLINE EXCLUSIVE PRICE
    141.00 €
    133.00 € / Pack of 96
    Save 8.00 €
    5% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit is used for post-sequencing clean-up in the MicroSEQ ID workflow.

The MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Refill Kit is used for post-sequencing clean-up in the MicroSEQ™ ID workflow. It includes the 8-well strips that are part of the MicroSEQ™ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246) but does not include the 96-well receiver plate. The gel filtration matrix used in the strip cartridges provides a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions.

The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit can be used downstream of the following sequencing kits:

  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit

The kit consists of twelve 8-well strips suitable for 96 sequencing samples. It is ideal for low- to medium-throughput microbial-identification workflows. The end products can be used directly for sequencing on any Applied Biosystems™ genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Sequencing Products
Format Strips
Content And Storage Sufficient for 96 sequencing samples

12 x 8-strips clean-up tubes
For Use With (Application) Sequencing
High-throughput Compatibility Multiplexing
Product Line MicroSEQ™ ID
Purification Target Fluorescent Dye Removal
Quantity 96 Samples
How do Phasemaker Tubes work?

Phasemaker Tubes work on the principle of density gradient centrifugation. Phasemaker Tubes contain a thick liquid polymer that is heavier than the aqueous phase of the TRIzol Reagent mix, but lighter than the organic phase, so after centrifugation, the polymer positions itself between these two layers and forms an exclusive physical barrier. It separates and completely isolates the upper aqueous phase from the organic phase underneath, making it very easy to aspirate the upper phase containing the RNA.

How long is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) stable for?

The product is guaranteed stable for one year when stored properly at 4 degrees C.

Can I store the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) at room temperature (20-24 degrees C)?

Yes. The kit contents are stable when stored in their original packaging at room temperature for up to 4 weeks. However, the recommended storage condition is at 4 degrees C.

How many sequencing samples is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256) sufficient for?

The MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit is sufficient for 96 sequencing samples.

What are the catalog numbers for the MicroSEQ ID Ultra Sequencing Clean-up Kits?

Here the kits with the catalog numbers:

- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246)
- MicroSEQ ID Ultra Sequencing Clean-up Plates Kit (Cat. No.A33245)
- MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)

What is included in the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)?

The kits consists of twelve 8-well strips suitable for 96 sequencing samples.

How long is the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips good for use once opened?

Strips should be used within 2 weeks of opening. We recommend returning unused strips from the 96-well plate holder to the foil package, sealing, and storing at 4 degrees C.

What is the minimum reaction volume for use of the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)?

The minimum reaction volume is 5 µL. The reaction volume should be brought up to at least 5 µL with distilled water before adding to the 8-well strips.

What is the storage condition for the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Refill Kit (Cat. No. A33256)?

The kit is stable when stored for 12 months at 4 degrees C or for 4 weeks at ambient temperature (20-24 degrees C).


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.