missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Ultra Sequencing Clean-up Plates Kit

Product Code. 18725515
Change view
Click to view available options
Quantity:
192 samples
Unit Size:
Pack of 192
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18725515 192 samples Pack of 192
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18725515 Supplier Applied Biosystems™ Supplier No. A33245

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MicroSEQ ID Ultra Sequencing Clean-up Plates Kit is used for post-sequencing clean-up in the MicroSEQ ID workflow.

The MicroSEQ™ ID Ultra Sequencing Clean-up Plates Kit is used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel-filtration matrix used in the microplate cartridges provides a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions. MicroSEQ ID Ultra Sequencing Clean-up plates can be stored at ambient temperate for up to 4 weeks. The MicroSEQ ID Ultra Sequencing Clean-up Plates Kit offers the same or better sequence quality results as the MicroSEQ™ ID Sequencing Clean-up Plates.

The MicroSEQ ID Ultra Sequencing Clean-up plates can be used downstream of the following sequencing kits:

  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit

This kit provides two 96-well microplate cartridges ideal for high-throughput microbial-identification workflows. The end products can be used directly for sequencing on any Applied Biosystems™ genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2 x 96-well reaction plates
• 2 x 96-well plate lids
• 2 x 96-well waste plates
• 2 x 96-well semi-skirted capillary plates

Format 96-well Plate
Sample Type Sequencing Products
For Use With (Application) Sequencing
High-throughput Compatibility High-throughput Compatible
No. of Samples 192 Samples
Product Line MicroSEQ™ ID
Quantity 192 samples
Target Fluorescent Dye Removal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.