missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Sequencing Clean-up Cartridges

Product Code. 30000677 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
36 cartridges
Unit Size:
Pack of 36
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000677 36 cartridges Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000677 Supplier Applied Biosystems™ Supplier No. 4408228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MicroSEQ ID Sequencing Clean-up Cartridges are used for post-sequencing clean-up in the MicroSEQ ID workflow.

The MicroSEQ™ ID Sequencing Clean-up Cartridges are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrix used in the cartridges provides a convenient and simple way to remove dye terminators, dNTPs, and salts, as well as residual primers from cycle sequencing reactions.

The MicroSEQ™ ID Sequencing Clean-up Cartridges can be used downstream of the following sequencing kits:

  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit

The kit consists of 36 cartridges and is ideal for low-throughput microbial identification workflows. The end products can be directly used for sequencing on any of Applied Biosystems™ genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Format Cartridge, Spin Column
Isolation Technology Spin Column
Final Product Type DNA
For Use With (Equipment) MicroSEQ™ ID
For Use With (Application) Sequencing
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line MicroSEQ
Quantity 36 cartridges
Purity Sequencing Grade
Shipping Condition Wet Ice
Target Sequencing Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.