missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroSEQ™ ID Sequencing Clean-up Plates

Product Code. 30000683 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 x 96-well plates
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000683 2 x 96-well plates Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000683 Supplier Applied Biosystems™ Supplier No. 4408227
  • ONLINE EXCLUSIVE PRICE
    369.00 €
    354.00 € / Pack of 2
    Save 15.00 €
    4% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The MicroSEQ ID Sequencing Clean-up Plates are used for post-sequencing clean-up in the MicroSEQ ID workflow.

The MicroSEQ™ ID Sequencing Clean-up Plates are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrix used in the microplate cartridges provides a convenient and simple way to remove dye terminators, dNTPs, and salts, as well as residual primers from cycle sequencing reactions.

The MicroSEQ™ ID Sequencing Clean-up Plates can be used downstream of the following sequencing kits:

  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit

This product provides two 96-well microplate cartridges ideal for high throughput microbial identification workflows. The end products can be directly used for sequencing on any Applied Biosystems™ genetic analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 x 96-well plates
For Use With (Equipment) 3500xL Genetic Analyzer, MicroSEQ™ ID, 3500 Genetic Analyzer
Product Type Microplate
Shipping Condition Wet Ice
Content And Storage Store in refrigerator (2°C to 8°C).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.