missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma with top quality media from the experts in media. Thermo Scientific™ MicroTest™ Tubes come in four media formulations to offer flexibility in both transport and storage.
Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.
MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae. MicroTest™ M5™ contains vancomycin, amphotericin B, and colistin and protein stabilizers for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest Tubes come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads.
Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.
Trusted reliable results
Not all products are available for sale in all territories.
Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae. MicroTest™ M5™ contains vancomycin, amphotericin B, and colistin and protein stabilizers for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.
MicroTest Tubes come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads.
Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.
Trusted reliable results
- Stringent quality control, including cellular toxicity on each lot number
- Patented formulations with stabilizing proteins, buffers, and specific antibiotics
- Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
- Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
- 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
- Individual transport media tubes or complete kits offer flexibility and versatility
- Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
- Kits include leak-resistant polypropylene bag for safe sample transport
Not all products are available for sale in all territories.
Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Product Type | Transport Tube |
| Format | Tube |
| Media Type | M4 |
| Closure Color | Blue |
| Compatibility | Culture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology |
| Description | M4 Tube |
| For Use With (Application) | Viruses: Chlamydiae, Ureaplasmas, and Mycoplasmas |
| Includes | Glass Beads |
| Packaging | Packaged in partitioned boxes. |
| Quantity | 72 Tubes/Pk. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?