Learn More
IBA Lifesciences Strep-Tactin™ Coated Microplate
Strep-Tactin™ coated microplate is a ready-to-use microplate for assay applications
Brand:  IBA Lifesciences 2-1501-001
Code : LD
Additional Details : Weight : 0.10000kg
Product Code. 12688247
  • 63.25 € / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Ready-to-use Strep-Tactin™-coated microplate (12 x 8-well strips) for ligand binding assays, immunoassays, enzyme assays, or diagnostic assays under high throughput conditions.
  • Due to the high specificity and affinity (µM-nM) of the streptavidin variant Strep-Tactin™, Strep-tag™II or Twin-Strep-tag™ fusion proteins can be selectively and stably captured even from complex mixtures.
  • Biomolecules of interest are presented to interaction partners in a uniform manner, resulting in reliable and reproducible assays.
  • The solid-phase, multi-well format is compatible with standard multichannel pipettes and automated plate readers.
  • Twin-Strep-tag™ fusion protein binding capacity determined with bacterial alkaline phosphatase (97 kDa) is 2 pmol for 200 µl incubation volume. 
Specifications
Specifications
Standard
96
300 μL
4°C
8 x 12
Strep-Tactin
1 Pk.