Learn More
Thermo Scientific™ Microtiter Tubes and Racks
Product Code. 11539954 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Product Type:
Frosted MicroTiter Tube
Microtiter Tube
Sterility:
Non-sterile
Sterile
Format:
Bulk
Robotic Rack
Unit Size:
Pack of 1000
Pack of 10000
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Sterility Format Unit Size
11539954 Microtiter Tube Non-sterile Bulk Pack of 1000
12358202 Frosted MicroTiter Tube Non-sterile Robotic Rack Pack of 960
11579944 Microtiter Tube Sterile Robotic Rack Pack of 960
11937675 Microtiter Tube Non-sterile Robotic Rack Pack of 960
10707994 Frosted MicroTiter Tube Non-sterile Bulk Pack of 10000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11539954 Supplier Thermo Scientific™ Supplier No. 3492
  • ONLINE EXCLUSIVE PRICE
    47.20 €
    39.10 € / Pack of 1000
    Save 8.10 €
    17% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Seal microtiter tubes tightly for storage with these polyethylene plug strips -- for use with 1.2mL MCT titertube racks.
Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.
Microtiter Tubes and Caps
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight-plug strips can be easily cut apart for individual samples
  • 1.2mL microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
  • Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
  • Microtiter tube system is available in bulk packaging or with a SBS standard footprint
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
  • Rigorous testing to ensure sample integrity and security
Packaging Options
  • Microtiter tubes and caps are available sterile and nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified free of RNase, DNase, DNA and pyrogens
Color Natural
Volume (Metric) 1.2 mL
Quantity 1000 Tubes/Unit, 10 Units/Cs.
Closure Material Polyethylene (Plug)
Product Line Thermo Scientific
Sterility Non-sterile
Format Bulk
Product Type Microtiter Tube