Learn More
Thermo Scientific™ Microtiter Tubes and Racks
Seal microtiter tubes tightly for storage with these polyethylene plug strips -- for use with 1.2mL MCT titertube racks.
Brand:  Thermo Scientific™ 3424-11
Code : A0
Additional Details : Weight : 1.14300kg
 View more versions of this product
Product Code. 12358202
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.
Microtiter Tubes and Caps
  • Polyethylene plug strip seals the Microtiter tubes tightly for storage
  • Eight plug strips can be easily cut apart for individual samples
  • 1.2mL Microtiter tubes have additional volume capacity for sample dilution and are ideal  for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
  • Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
  • Microtiter tube system is available in bulk packaging or with a SBS standard footprint
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
  • Rigorous testing to ensure sample integrity and security
Packaging Options
  • Microtiter tubes and caps are available sterile and nonsterile
Specifications
Specifications
Certified free of RNase, DNase, DNA and pyrogens
1.2mL Frosted MicroTiter Tube Robotic Rack, Nonsterile
96 tubes/rack 10 racks/unit, 5 units case
Non-sterile
Microtiter
Natural
1.2 mL
Thermo Scientific™
Polyethylene (Plug)