missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Microtube Floating Storage Racks

Product Code. 17547896
Click to view available options
Size:
24-place
48-place
Closure Included:
Clear Lid
None
Unit Size:
Pack of 10
Pack of 5
This item is not returnable. View return policy

Product Code. 17547896

Brand: Labcon™ 30362200009

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Available in 24- or 48-place arrays with or without lids.

  • Holds 1.5, 1.7, or 2 mL microtubes
  • Freezable to -80°C
  • Made in USA

Specifications

Closure Included Clear Lid
Autoclavable Autoclavable
Color White
Holds 1.5, 1.7, 2 mL Microtubes
Material Polypropylene
Size 24-place
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.