missing translation for 'onlineSavingsMsg'
Learn More

Midecamycin, 900μg/mg, Thermo Scientific™

A broad spectrum antibiotic

Brand:  Thermo Scientific Alfa Aesar J66046.03

Quantity 1g

 View more versions of this product

Product Code. 15477249

  • 222.00 € / 1g
 Restricted Item
Estimated Shipment: 08-10-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

35457-80-8
813.979
DMUAPQTXSSNEDD-RBATUFKESA-N
[(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
C41H67NO15
MFCD00865000
124225192
CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C

Specifications

35457-80-8
1g
C41H67NO15
874°C
DMUAPQTXSSNEDD-RBATUFKESA-N
[(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
124225192
Midecamycin, 900μg/mg
White
>110°C (230°F)
MFCD00865000
Soluble in methanol or ethanol; Insoluble in water
CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C
813.979
813.97
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.