missing translation for 'onlineSavingsMsg'
Learn More

Midecamycin, TRC

Product Code. 17914405
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914405 1 g 1g
17971475 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17914405 Supplier Toronto Research Chemicals Supplier No. TRCM3432501G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35457-80-8
Molecular Formula C41 H67 N O15
Molecular Weight (g/mol) 813.97
Synonym Leucomycin V, 3,4B-dipropanoate, Oxacyclohexadecane, leucomycin V deriv., Aboren, Antibiotic SF 837, Antibiotic SF 837A1, Antibiotic YL 704B1, Espinomycin A, Macropen, Medecamycin A1, Medemycin, Medemycin A1, Midecamycin, Midecamycin A1, Midecin, Momicine, Mydecamycin, Mydecamycin A1, Myoxam, NSC 154011, Normicina, Platenomycin B1, Rubimycin, SF 837, SF 837A1, Turimycin P3, YL 704B1
IUPAC Name [(4R,5S,6S,7R,9R,10R,11E,13E,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-4-yl] propanoate
SMILES CCC(=O)O[C@@H]1CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@H](O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](OC(=O)CC)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@H]1OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C41H67NO15/c1-11-30(45)54-29-21-32(47)51-24(4)16-14-13-15-17-28(44)23(3)20-27(18-19-43)37(38(29)50-10)57-40-35(48)34(42(8)9)36(25(5)53-40)56-33-22-41(7,49)39(26(6)52-33)55-31(46)12-2/h13-15,17,19,23-29,33-40,44,48-49H,11-12,16,18,20-22H2,1-10H3/b14-13+,17-15+/t23-,24-,25-,26+,27+,28+,29-,33+,34-,35-,36-,37+,38+,39+,40+,41-/m1/s1
Formula Weight 813.4511
Chemical Name or Material Midecamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.