Learn More
Miele Professional™ Miele Process Documentation
Product Code. 15480445 Shop All Miele Products
Click to view available options
:
with Miele G7831, G 788x ,G 789x, and PS 120xB; model: Net500.
Product Code. 15480445
Brand: Miele Professional™ 7951420
This item requires a price update prior to being ordered.
Request a Price for this item here.
APH 301 Connection cable type 1 for connection of a machine with serial interface for a PC/Net500.
  • Compatible with Miele G7831, G 788x ,G 789x, and PS 120xB
  • Compatible with numerous appliances from Melag, Sirona, W and H and Hawo
  • Cable length 3 m
TRUSTED_SUSTAINABILITY
Specifications
Product Type Process/Operating Documentation/Communication
For Use With (Equipment) Washer-disinfectors, laboratory