missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Cable category 6
- Color: Transparent silver
- Used for the connection of Miele machines
- Used for the network connection of Melag sterilizers
Specifications
Specifications
| Product Type | Process/Operating Documentation/Communication |
| For Use With (Equipment) | Washer-disinfectors, laboratory |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?