missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ MIG FIT Welder Gloves
Description
- Cowhide split leather glove
- Gunn cut, winged thumb, gauntlet type
- Aluminised Kevlar™ back of the hand with cotton fleece liner
- Heat resistant split leather palm with cotton fleece lining
- Split leather cuff with cotton fleece liner on palm side
- Aluminised Kevlar™ cuff with cotton fleece liner on back side
- Thickness leather only 1.1/1.3 mm
- Split leather is an ideal material for the production of welding gloves
- Winged thumb and the flexible materials used make this glove ergonomic, thus providing the user with comfort of use
- The cotton fleece lining increases the glove's thermal insulation
- The aluminised Kevlar™ textile back offers strong resistance to heat, radiation and small splashes of molten metal
- Its cotton fleece reinforcement inside the glove and at the side of the hand provide good support on the hot metal part
- Wear by abrasion is limited due to the leather reinforcement at the thumb seam
- The split leather cuff helps to protect a large part of the forearm against mechanical hazards
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY : II Intermediary risks In accordance with standards: EN 420-2003 Basic standards EN 388-2003 Mechanical Hazards EN 407-2004 Heat hazards EN 12 477-2000 type A Protection for Welders |
| Cuff Material | Dupont™ Kevlar™ aramid fibers |
| Color | White, Orange |
| Material | Cowhide Leather |
| Size | 10 |
| Thickness (Metric) | 1.1 to 1.3 mm |
| Liner Material | Cotton |
| Length (Metric) | 35.5 cm |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?