missing translation for 'onlineSavingsMsg'
Learn More

Hoefer Mighty Small precast gels

Product Code. 30286592
Change view
Click to view available options
Gel Percentage:
10%
12%
4 to 12%
4 to 20%
8%
Wells:
12-well
15-well
Unit Size:
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Percentage Wells Unit Size
30286592 10% 12-well Pack of 10
30286587 8% 15-well Pack of 10
30286582 8% 12-well Pack of 10
30286584 10% 15-well Pack of 10
30286589 12% 15-well Pack of 10
30286586 12% 12-well Pack of 10
30286588 4 to 12% 15-well Pack of 10
30286590 4 to 12% 12-well Pack of 10
30286583 4 to 20% 15-well Pack of 10
30286591 4 to 20% 12-well Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 30286592 Supplier Hoefer Supplier No. MSG010T
  • ONLINE EXCLUSIVE PRICE
    79.45 €
    59.13 € / Pack of 10
    Save 20.32 €
    25% Off
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hoefer Mighty Small precast gels are high-performance mini polyacrylamide gels commonly used for protein separation.

  • Specification:
  • Plate Size: 10 x 8 cm
  • Gel Size: 8.5 x 7 cm, 1 mm thickness
  • Wells: 15 well (12 wells available)
  • System/Buffer: Bis-Tris System, MOPS Buffer
  • Loading Volume: For 15 wells: max 30 μl (recommended: ≤ 15 μl)
  • Package: 10 pcs/box
  • Weight:0.66 lb.
  • Storage temp: 2 to 8°
  • Shelf life: 12 months
  • Advantages:
  • upgraded Bis-tris system with moderate PH to ensure less modification on the protein to get a sharp and clear band
  • Save trouble and time from making gels

Specifications

Buffer MOPS
Color Clear
Concentration 10%
Gel Percentage 10%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
pH Range 7
Sample Loading Volume 25 μL
Separation of Protein
Separation Range 10 to 300 kDa
Separation Type Denaturing
Wells 12-well
Quantity 10
Product Type Precast Gel
For Use With (Equipment) Mini Gel Tank
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.