missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer Mighty Small precast gels
Click to view available options
Gel Percentage:
10%
12%
4 to 12%
4 to 20%
8%
Wells:
12-well
15-well
Unit Size:
Pack of 10
Description
- Specification:
- Plate Size: 10 x 8 cm
- Gel Size: 8.5 x 7 cm, 1 mm thickness
- Wells: 15 well (12 wells available)
- System/Buffer: Bis-Tris System, MOPS Buffer
- Loading Volume: For 15 wells: max 30 μl (recommended: ≤ 15 μl)
- Package: 10 pcs/box
- Weight:0.66 lb.
- Storage temp: 2 to 8°
- Shelf life: 12 months
- Advantages:
- upgraded Bis-tris system with moderate PH to ensure less modification on the protein to get a sharp and clear band
- Save trouble and time from making gels
Specifications
Specifications
| Buffer | MOPS |
| Color | Clear |
| Concentration | 4 to 20% |
| For Use With (Equipment) | Mini Gel Tank |
| Gel Percentage | 4 to 20% |
| Gel Size | Mini |
| Gel Thickness | 1.0 mm |
| Gel Type | Bis-Tris |
| pH Range | 7 |
| Product Type | Precast gels |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction