missing translation for 'onlineSavingsMsg'
Learn More

Hoefer Mighty Small precast gels

Product Code. 30286590
Click to view available options
Gel Percentage:
10%
12%
4 to 12%
4 to 20%
8%
Wells:
12-well
15-well
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 30286590

Brand: Hoefer MSG412T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hoefer Mighty Small precast gels are high-performance mini polyacrylamide gels commonly used for protein separation.

  • Specification:
  • Plate Size: 10 x 8 cm
  • Gel Size: 8.5 x 7 cm, 1 mm thickness
  • Wells: 15 well (12 wells available)
  • System/Buffer: Bis-Tris System, MOPS Buffer
  • Loading Volume: For 15 wells: max 30 μl (recommended: ≤ 15 μl)
  • Package: 10 pcs/box
  • Weight:0.66 lb.
  • Storage temp: 2 to 8°
  • Shelf life: 12 months
  • Advantages:
  • upgraded Bis-tris system with moderate PH to ensure less modification on the protein to get a sharp and clear band
  • Save trouble and time from making gels

Specifications

Buffer MOPS
Color Clear
Concentration 4 to 12%
For Use With (Equipment) Mini Gel Tank
Gel Percentage 4 to 12%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Bis-Tris
pH Range 7
Product Type Precast gels
Sample Loading Volume 25 μL
Separation of Protein
Separation Range 10 to 300 kDa
Separation Type Denaturing
Wells 12-well
Quantity 10
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.