Learn More
Emma Safety Footwear Mike XL Safety Shoes
Product Code. 15978121
Change view
Click to view available options
Size:
50
51
52
53
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15978121 50 1 pair
15988121 51 1 pair
15998121 52 1 pair
15908131 53 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15978121 Supplier Emma Safety Footwear Supplier No. MM10035850
  • ONLINE EXCLUSIVE PRICE
    277.20 €
    173.25 € / 1 pairs
    Save 103.95 €
    37% Off
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
Ideal for working with metal with optimal comfort, ventilation and moisture removal technology. A perfectly integrated steel toe protects feet against falling pieces of metal and other exterior threats. The Gripforce™ DUO soles withstand high temperatures and are oil and petrol resistant.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Mike XL
Liner Material Full grain Nappa leather / Split leather
Material Leather
Size 50
Toe Material Steel