missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mildronate dihydrate

Product Code. 16677062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16677062 1 g 1g
16687062 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16677062 Supplier Thermo Scientific Chemicals Supplier No. 466260010
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86426-17-7
Molecular Formula C6 H14 N2 O2 . 2 H2 O

Specifications

Quantity 1 g
Solubility soluble aq. soln.
Formula Weight 182.22
Physical Form Crystalline Powder
Product Type Reagent
Purity 97.5 to 102.5% (Titration with HClO4)
InChI Key JFWLFLLRLZSBRA-UHFFFAOYSA-N
SMILES O.O.C[N+](C)(C)NCCC([O-])=O
IUPAC Name 3-(2,2,2-trimethylhydrazin-2-ium-1-yl)propanoate dihydrate
Molecular Weight (g/mol) 182.22
Chemical Name or Material Mildronate dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.