missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Milk Plate Count Agar (Dehydrated)

Product Code. 10750944
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.50kg
500g
This item is not returnable. View return policy

Product Code. 10750944

Brand: Thermo Scientific™ CM0681B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and enumerate microorganisms in milk and dairy products with this medium while conforming to ISO standards.

Isolate and enumerate microorganisms in milk and dairy products with Thermo Scientific™ Oxoid™ Milk Plate Count Agar (Dehydrated). Milk Plate Count Agar is equivalent to the medium recommended by British Standard Institution1 and International Organization for Standardization2.

Liquid milk and other dairy products are highly perishable and can spoil in a few days. The bacteria present in dairy products may cause disease or spoilage. Foodborne illness from dairy product consumption of have been predominantly associated with Salmonella enterica, Listeria monocytogenes, Camplylobacter jejuni and Escherichia coli O157: H7.

Milk Plate Count Agar is recommended for the enumeration of microorganisms in liquid milk, ice cream, dried milk and whey. Tryptone in the medium provides amino acids and other complex nitrogenous substances. Yeast extract supplies vitamins. Addition of skim milk in the medium makes the conditions optimal for microorganisms which grow in milk.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: NMKL standards.
Description Milk Plate Count Agar (w/antibiotic-free skimmed milk)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 25.6L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.