missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Millenia Safety Glasses

Product Code. 12642743
Change view
Click to view available options
Clear lens, black frame
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12642743 Clear lens, black frame
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12642743 Supplier Honeywell™ Supplier No. 1002781
  • ONLINE EXCLUSIVE PRICE
    28.74 €
    12.09 € / Pack of 1
    Save 16.65 €
    57% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Balanced, light frame for fit, comfort, and protection.

  • For general use and protection against impact (45 m/s)
  • Suitable for mechanical work: grinding, turning, milling, etc.
  • Polycarbonate lenses offer 99.9% UV protection up to 380 nm
  • Protection against fogging
  • Balanced fit
  • Adjustable, breakaway Flexicord retainer
  • Unique 9-base curved lens design
  • Excellent optical qualities
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: EC 1131, EC Attestation: EC 131
Lens Coating Anti-fog
Lens Tint Clear
For Use With (Application) General use and protection against impact (45m/s), mechanical work such as grinding, turning, milling
Frame Color Black
Includes Breakaway Flexicord retainer
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.