missing translation for 'onlineSavingsMsg'
Learn More

Melford Millers LB Broth Granulated

Product Code. 30018429
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018429 500 g 500g
30018428 1 kg 1kg
30018430 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018429 Supplier Melford Supplier No. L24400500.0
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For propagation and maintenance of E.

For propagation and maintenance of E. Coli. High Salt Formula. Molecular Biology Grade.

Dissolve 25 grams in 1 liter distilled water and adjust the pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes. Material specially granulated to reduce dust and increase solubility rate.

Ingredients:

Tryptone:10 g/l

Sodium Chloride: 10 g/l

Yeast Extract: 5 g/l

Specifications

Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.