Learn More
MilliporeSigma Millicell™ Disposable Hemocytometer
Product Code. 18854582 Shop All Merck Products
Change view
Click to view available options
Includes:
2 Chambers/Slide
Unit Size:
Pack of 50
Product Code. Includes Unit Size
18854582 2 Chambers/Slide Pack of 50
1 options
Product Code. 18854582 Supplier MilliporeSigma Supplier No. MDH2N150PK
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Individually wrapped Neubauer Improved hemocytometer for counting cells.
The Millicell™ disposable hemocytometer is used to manually count cells. It consists of a surface-patterned two or four enclosed chambers with ports for sample injection. Count cells in the same way as you do with the Neubauer Improved counting chamber.
The Neubauer Improved counting chamber consists of 9 large squares, each measuring 1 x 1 mm. Each square has a total volume of 0.1 mm3. The cells are distributed over 3 x 3 large squares, each with 1 mm edge length and with a surface area of 1 mm2.
With the Millicell™ disposable hemocytometer, you inject 10 μL of sample, stained or unstained, into the desired chamber.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hemacytometer
Grid Type Neubauer Improved
For Use With (Application) Cell counting
Includes 2 Chambers/Slide