missing translation for 'onlineSavingsMsg'
Learn More

Merck Millicell Hanging Cell Culture Insert

Product Code. 15515960 Shop All Merck Products
Change view
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
8 μm
No. of Wells:
6
12
24
Unit Size:
Pack of 48
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size No. of Wells unitSize
15515960 0.4 μm 6 Pack of 48
15565960 1 μm 6 Pack of 48
15585960 3 μm 6 Pack of 48
15545960 5 μm 6 Pack of 48
15525960 0.4 μm 12 Pack of 48
15303328 1 μm 12 Pack of 48
15392839 3 μm 12 Pack of 48
15595950 8 μm 12 Pack of 48
15535960 0.4 μm 24 Pack of 48
15449779 1 μm 24 Pack of 48
15505970 3 μm 24 Pack of 48
15431055 5 μm 24 Pack of 48
15505960 8 μm 24 Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Product Code. 15515960 Supplier Merck Supplier No. MCHT06H48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allows attachment or suspension cells to access media from both their apical and basolateral sides.

  • Cell growth, structure, and function more closely mimic what occurs in vivo
  • In addition, Millicell inserts make it possible to study both sides of the cell monolayer
  • The inserts are easily prepared for SEM and TEM visualizing techniques, and they are compatible with cellular and/or fluorescent stains
  • For co-culturing and permeability assays
  • Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
  • Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy
  • PET Membrane (polyethylene terephthalate) for growth of attachment-dependent cells without matrix
  • Individually blister packaged
TRUSTED_SUSTAINABILITY

Specifications

Description PET (Polyethylene Terephthalate) Filter Code
Material Polystyrene
Pore Size 0.4 μm
Type Cell Culture Insert
Item Description 50°C Max. Operating Temperature
For Use With (Application) Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry
Diameter (Metric) Filter 24 mm
Quantity 48/Pk
For Use With 6-well Plates
Color Clear
Sterility Sterile
No. of Wells 6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.