missing translation for 'onlineSavingsMsg'
Learn More

Merck MilliSolve Kit

Product Code. 15804176
Change view
Click to view available options
Diameter (Metric) Inner:
12.5 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
15804176 12.5 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15804176 Supplier Merck Supplier No. XX1614700
  • ONLINE EXCLUSIVE PRICE
    1750.00 €
    1598.25 € / Pack of 1
    Save 151.75 €
    8% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Millipore MilliSolve Filtration System has been designed for the filtration of liquid chromatography buffers and solvents under vacuum.

Millipore MilliSolve Filtration System - Kit

The Millipore MilliSolve Filtration System has been designed for the filtration of liquid chromatography buffers and solvents under vacuum. The system uses a 0.45 μm membrane filter to eliminate particles which can shorten column life. Vacuum filtration with the Millipore MilliSolve system also removes a large portion of dissolved gasses from buffers, reducing the risk of air bubbles interfering with LC instruments. With automatic and continuous filtration, there is no need to add liquid into a funnel as filtration progresses. Filtration occurs in a closed system, which is important when filtering hazardous fluids. If open funnel filtration is required, the 300 mL funnel replaces the vacuum cover.
This item is a kit composed of the following items:

XX1614705 - Millipore Ground Joint Flask, 2L, Conical bottom
XX1514702 - Millipore Glass Base and Cap, 47 mm, Glass frit support
XX1614701 - Millipore Glass cover, 47 mm, For use with Millipore MilliSolve Kit
XX1014703 - Millipore Spring Clamp, 47 mm, Aluminum
XT1200000 - Millipore Tubing for Solvent/Buffer, PTFE, 70 cm
XX1014704 - Millipore Glass funnel, 300 mL, 47 mm, Borosilicate, Ground glass seal
ALIGNED05 - Millipore Alignment Guide, 47 mm, 5/PK

 

Features & Benefits:

Automatic, continuous solvent and buffer filtration eliminates the need to pour during filtration

  • 2L receiving flask includes plug and tubing for storage, eliminating filtered buffer transfer, reducing labor and lessening contamination risk
  • Conical 2L flask bottom allows access to all filtered solvent or buffer

Applications: Mobile Phase Preparation, Buffer Filtration

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 12.5 cm
Includes 2 L Flask, 47 mm dia. filter, Borosilicate Glass, Anodized Aluminum clamp, 120 in. PTFE tubing, Stainless Steel forceps, Glass vacuum support base with tube cap
Product Type Laboratory Filter Holders or Cyclones
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.