missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Milnacipran hydrochloride

Product Code. 16657072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16657072 1 g 1g
16667072 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16657072 Fornitore Thermo Scientific Chemicals N. del fornitore 466570010
 Restricted Item
Estimated Shipment: 30-07-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 101152-94-7
Molecular Formula C15 H22 N2 O . H Cl

Specifica

Quantity 1 g
Solubility soluble aq. soln.
Formula Weight 282.81
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
InChI Key XNCDYJFPRPDERF-PBCQUBLHSA-N
SMILES [H+].[Cl-].CCN(CC)C(=O)[C@@]1(C[C@@H]1CN)C1=CC=CC=C1
IUPAC Name hydrogen (1R,2S)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide chloride
Molecular Weight (g/mol) 282.81
Chemical Name or Material Milnacipran hydrochloride
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.