missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil (Heavy), TRC

Product Code. 30479627
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479627 25 mL 25mL
30470056 5 mL 5mL
30470849 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479627 Supplier Toronto Research Chemicals Supplier No. TRCM34495025ML
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5

Specifications

Quantity 25 mL
Synonym White mineral oil (petroleum), Mineral Oil Heavy, Mineral Oil
IUPAC Name White mineral oil
Chemical Name or Material Mineral Oil (without additives)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.