missing translation for 'onlineSavingsMsg'
Learn More

Mineral oil, MP Biomedicals™

Product Code. 11462442 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462442 1 L 1 L
11416562 100 mL 100mL
11452442 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462442 Supplier MP Biomedicals Supplier No. 0215169491
  • ONLINE EXCLUSIVE PRICE
    55.35 €
    52.50 € / 1 L
    Save 2.85 €
    5% Off
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Density 0.81 to 0.894 at 15°C
Quantity 1 L
Physical Form Viscous Liquid
Chemical Name or Material Mineral oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.