missing translation for 'onlineSavingsMsg'
Learn More

Mineral Oil, MP Biomedicals™

Product Code. 11426562 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426562 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11426562 Supplier MP Biomedicals Supplier No. 0215013890
  • ONLINE EXCLUSIVE PRICE
    142.00 €
    134.40 € / 500mL
    Save 7.60 €
    5% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as an overlay covering microdrops of culture medium in petri dishes, during the culture of oocytes and embryos in IVF and related procedures. Mineral Oil, MP Biomedicals is a transparent, colorless and odorless oil, a liquid by-product of the distill

A mineral oil is any of various colorless, odorless, light mixtures of alkanes in the C15 to C40 range from a non-vegetable (mineral) source. Most often, mineral oil is a liquid by-product of the distillation of petroleum to produce gasoline and other petroleum-based products from crude oil. A mineral oil in this sense is composed mainly of alkanes and cyclic paraffins, related to petroleum jelly (also known as “white petrolatum”). It has applications in laboratory applications and also in food processing, cosmetics and pharmaceuticals.

Specifications

CAS 8042-47-5
Density 0.81–0.894 at 15°C
Boiling Point 218°C to 643°C (1013 hPa)
Molecular Formula C16H10N2Na2O7S2
Refractive Index n20/D 1.467 (Lit.)
Quantity 500 mL
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
Solubility Information Insoluble in water.
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
Molecular Weight (g/mol) 452.363
PubChem CID 9566064
Viscosity 65.80–71.00 Centistokes (at 40°C) (Lit.)
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.