missing translation for 'onlineSavingsMsg'
Learn More

Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.

Product Code. 10458470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
6 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
6mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10458470 2.5 L Glass Bottle 2.5 L
10468470 6 mL Glass Bottle 6mL
10523481 1 L Glass Bottle 1 L
10135543 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10458470 Supplier Thermo Scientific Acros Supplier No. 124020025
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For IR spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00131611
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Density 0.8510g/mL
Flash Point 185°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4620 to 1.4730
Quantity 2.5 L
Merck Index 14, 7186
Specific Gravity 0.851
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether,, carbon dislulfide, petroleum ether, oils, miscible with wax and fats, insoluble in alcohol
Viscosity 14-19 mm2/s (40°C)
Grade Spectroscopy
Physical Form Oily Liquid
Chemical Name or Material Mineral oil
Show More Show Less
compliance-icons
Product Identifier
  • Mineral oil, suitable for nujol mull preparations for IR-spect.
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.