Learn More
Cytiva Mini Dialysis Kits
Product Code. 11510694 Shop All Cytiva Products
Change view
Click to view available options
MWCO:
1 kDa
8 kDa
Capacity (Metric):
250 μL
2 mL
Unit Size:
1 set
250µL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Capacity (Metric) Unit Size
11510694 1 kDa 2 mL Each
11520694 8 kDa 2 mL 1 set
10196074 1 kDa 250 μL 1 set
10617465 8 kDa 250 μL 250µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11510694 Supplier Cytiva Supplier No. 80648394
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Includes
50 tubes with 50 dialysis and 50 standard caps, plus six floats
Offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
  • Especially well-suited for desalting protein samples prior to analysis by isoelectric focusing (IEF) and 2-D electrophoresis
  • Uses disposable dialysis tubes with dialysis membrane incorporated into caps
  • Conical tube bottom maximizes sample recovery
  • Process complete in as little as several hours
  • Choice of molecular weight cut-off and tube size
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mini Dialysis Kit
MWCO 1 kDa
Capacity (Metric) 2 mL
Kit Contents 50 x tube with dialysis cap, 50 x standard cap, 6 x float
Quantity Each
For Use With (Application) Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane