missing translation for 'onlineSavingsMsg'
Learn More

Tork Mini Toilet Seat Cleaner S5

Product Code. 30243769 Shop All Tork Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30243769 Clear Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30243769 Supplier Tork Supplier No. 425302
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tork Toilet Seat Cleaner provides extra reassurance to visitors. Ensure a cleaner toilet seat with one dose of Toilet Seat Cleaner on a piece of toilet paper. Suitable for Tork Mini Soap and Sanitizer Dispensers, proven easy to use.

  • Fast refilling in less than 10 seconds.
  • Hygienic refilling with a factory-sealed bottle with a new pump for every refill, which helps reduce the risk of contamination.
  • Reduce waste with this collapsible refill bottle, which leads to 70% less volume waste (based on Essity test).
  • For hazard and precaution information, please refer to the Safety Data Sheet.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Toilet Cleaner
Color-coded No
Scent Unscented
Color Clear
Packaging Plastic Bottle
Alternate Part Number 7322541450858
compliance-icons
Product Identifier
  • X8 Tork Mini Toilet Seat Cleaner S5 Factory-Sealed
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.