Learn More
Ekastu Safety™ Eye Wash Bottle with Funnel
Product Code. 10385722
Change view
Click to view available options
Product Type:
Eyewash Bottle
Eyewash Bottle, Empty
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
10385722 Eyewash Bottle, Empty Each
10470903 Eyewash Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10385722 Supplier Ekastu Safety™ Supplier No. 377951
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Made for immediate rinsing of the eyes after accidental splashing from acids, bases and other hazardous chemicals. EKASTU Safety™ Eye Wash Bottle with Funnel can be used in either upright or horizontal positions.
  • Eye Wash Bottle with funnel
  • DIN EN 15154-4
  • Supplied empty or filled
  • For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
  • PE bottle with screwed-on eye funnel and tilting valve
  • Can be used in both upright and horizontal positions
  • leak-proof
  • Stores easily in EPS Eye Wash Station (sold separately)
Specifications
Capacity (Metric) 600 mL
For Use With (Application) For Immediate Rinsing of the Eyes after Contact with Acids, Bases
Includes Screwed-on Eye Funnel and Tilting Cap
Product Type Eyewash Bottle, Empty