missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Miniature Breaker Lockout Demo Kit

Product Code. 15603547 Shop All Brady Products
Change view
Click to view available options
Color:
Black/Red
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15603547 Black/Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15603547 Supplier Brady™ Supplier No. 805853
  • ONLINE EXCLUSIVE PRICE
    73.75 €
    70.58 € / Pack of 1
    Save 3.17 €
    4% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains a variety of Minaiture Circuit Breaker Lockouts with a variety of Pin-outs.

  • Black and Red
  • Contains:
    • 1 Miniature Circuit Breaker Lockouts - Pin-in Standard,
    • 1 Miniature Circuit Breaker Lockouts - Pin-out Standard
    • 1 Miniature Circuit Breaker Lockouts - Pin-out Wide
    • 1 Miniature Circuit Breaker Lockouts - Tie Bar
    • 1 Miniature Circuit Breaker - Pin-out Wide (ABB-MS325)
    • 1 Universal Mini Circuit Breaker Lockout

Specifications

Color Black/Red
Diameter (Metric) Shackle 7 mm
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.