missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments Minicentrifuge-Vortex for PCR plates

Product Code. 30274349
Change view
Click to view available options
Capacity:
2 x 96-well PCR Plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
30274349 2 x 96-well PCR Plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30274349 Supplier Grant Instruments Supplier No. MSC2P
  • ONLINE EXCLUSIVE PRICE
    770.00 €
    762.00 € / Pack of 1
    Save 8.00 €
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Compact sized digital centrifuge intended to collect droplets, mix reagents and collect once more for improved PCR yield

  • The combination of spin-mix functions ensures fast operation, thorough mixing and repeatable results.
  • The centrifuge rotor can accommodate 2 unskirted or semi-skirted PCR plates

Possible to operate in 4 independent modes:

  • Centrifuge - Max. 3500 RPM
  • Vortex - up to 5 min.
  • Centrifuge/Vortex - combined two motion types
  • Spin-mix-spin algorithm - up to 10 cycles

The spin-mix-spin algorithm (SMS-algorithm) is designed to collect (or reset) micro volumes of reagents to the bottom of the PCR plate tubes (the first centrifugation or spin), then vortexing (mix) and re-collecting reagents (repeated spin) from the walls and cover.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 2 x 96-well PCR Plates
Max. RCF 2,500 x g
Max. Speed 3,500 rpm
Refrigerated No
Amperage 2 A
Height (English) Exterior 9.05 in.
Height (Metric) Exterior 230 mm
Width (English) Exterior 6.5 in.
Height (English) Exterior Lid Closed 9.05 in.
Height (English) Exterior Lid Open 14.2 in.
Height (Metric) Exterior Lid Closed 230 mm
Height (Metric) Exterior Lid Open 360 mm
Certifications/Compliance CE
Dimensions (L x W x H) Exterior 9.05 x 8.66 x 6.5 in.
Standards CE
Warranty 2 Years
Other Functions Centrifuge, Vortex, Spin-mix
Dimensions (HWD) 6.5 x 8.66 x 9.05 in.
For Use With (Equipment) 2 x 96-well PCR Plates
Net Weight 2.7 kg
Frequency 50/60 Hz
Voltage 100/240 V
Run Time 0 to 10 min.
Dimensions (H x W x D) 165 x 220 x 230 mm
Quantity 1 Ea.
Display Type LCD
Product Type Centrifuge Vortexer
Width (Metric) Exterior 165 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.