Learn More
HUBER Minichiller 600
For many laboratory applications. Huber Kaeltmaschinenbau Minichiller 600 is suitable for cooling reactor blocks, vacuum pumps, rotary evaporators or heat exchangers.
Brand:  HUBER 3066.0010.99
Product Code. 15554669
  • POA / Pack of 1
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
  • Environmentally-friendly, economic cooling solution
  • High efficiency
  • Precise temperature control
  • Powerful circulation pump
  • Robust stainless construction
  • Reliable continuously operation with warning functions
  • Easy to fill and empty
  • Illuminated filling level indicator
  • LED temperature display
  • Easy operation with only three keys