missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MiniCollect™ Tube. Lavender Cap, K3E K3EDTA, Non-Sterile

Product Code. 15748169
Change view
Click to view available options
Capacity (Metric):
0.25, 0.5 mL
1 mL
Unit Size:
Pack of 2000
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15748169 0.25, 0.5 mL Pack of 500
15830146 0.25, 0.5 mL Pack of 2000
15758169 1 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15748169 Supplier Greiner Bio-One Supplier No. 450530
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Inner wall is coated with anticoagulant that binds calcium ions and interrupts clotting.

  • Inner wall is coated with spray-dried K3E K3EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.
  • Used for hematology.
  • Available in different quantities.
TRUSTED_SUSTAINABILITY

Specifications

Additive K3 E K3 EDTA
Sterility Non-sterile
Capacity (Metric) 0.25, 0.5 mL
Closure Included Yes
Closure Color Lavender
compliance-icons
Product Identifier
  • X500 MiniCollect TUBE 0.25/0.5 ml K3E K3EDTA lave nder cap
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethylenediaminetetraacetic Acid Tripotassium Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.