missing translation for 'onlineSavingsMsg'
Learn More

Alpha Packaging™ Minigrip™ Polyethylene Resealable Bags

Product Code. 12380529
Change view
Click to view available options
Width (Metric):
150 mm
380 mm
Height (Metric):
140 mm
510 mm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) unitSize
12380529 150 mm 140 mm Pack of 100
12390549 380 mm 510 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12380529 Supplier Alpha Packaging™ Supplier No. 12380529
  • ONLINE EXCLUSIVE PRICE
    8.95 €
    7.15 € / Pack of 100
    Save 1.80 €
    20% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

Manufactured from polyethylene. Alpha Packaging™ Minigrip™ Polyethylene Resealable Bags are ideal for sending samples.

  • Resealable section for the specimen
  • Plain bag; no biohazard warning label
  • Opening along width
  • 50μm thickness.
  • Polyethylene
  • Three write-on bars for labelling

Specifications

Description Plain bag; no biohazard label
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Quantity 100 Pack
Height (Metric) 140 mm
Product Type Resealable Bag
Width (Metric) 150 mm
Thickness (Metric) 50 μm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.