Learn More
Lovibond™ Water Testing Minikit
Click to view available options
Range:
0 to 500 mg/L
0.75 to 10% acid
10 to 500 mg/L
2 to 50 mg/L
20 to 200 mg/L
20 to 500 mg/L
5 to 5000 mg/L
70 to 1500 mg/L
Analytes:
Alkalinity Total M
Alkalinity-P, CaC03
Chloride
Cleaning Acid Strength
Cyanuric Acid
Nitrite
Sulphate (low range) Na2SO4
Sulphite (high range)
Sulphite (low range)
Unit Size:
Each
Beschreibung
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
- Easy operation and exact reagent dosing
- High accuracy
- Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
- Unrestricted shipment
- Safe storage
Spezifikation
Spezifikation
| Analytes | Cleaning Acid Strength |
| Includes | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. |
| Range | 0.75 to 10% acid |
| Sufficient For | Approximately 40 Tests |
| Test Method | Tablet count |
| Type | Minikit water testing kit |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur