Learn More
Minoxidil
Description
Minoxidil is used as antihypertensive, antialopecia agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
- Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure
Specifications
Specifications
| Melting Point | 248°C to 265°C |
| Color | White |
| Quantity | 5 g |
| Merck Index | 14,6203 |
| Solubility Information | Soluble in water (2.2mg/ml),100% ethanol (29mg/ml),propylene glycol,acetone,DMSO (6.5mg/ml),and methanol. |
| Formula Weight | 209.25 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Minoxidil |
Safety and Handling
- Minoxidil
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Minoxidil
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.