missing translation for 'onlineSavingsMsg'
Learn More

MIR96-IN-1, MedChemExpress

Product Code. p-200101615
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16451559

Brand: Medchem Express HY15843

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MIR96-IN-1 targets the Drosha site in the miR-96 (miRNA-96, microRNA-96) hairpin precursor, inhibiting its biogenesis, derepressing downstream targets, and triggering apoptosis in breast cancer cells. MIR96-IN-1 binds to RNAs with Kds of 1.3, 9.4, 3.4, 1.3 and 7.4 μM for RNA1, RNA2, RNA3, RNA4 and RNA5, respectively.

MIR96-IN-1 targets the Drosha site in the miR-96 (miRNA-96, microRNA-96) hairpin precursor, inhibiting its biogenesis, derepressing downstream targets, and triggering apoptosis in breast cancer cells. MIR96-IN-1 binds to RNAs with Kds of 1.3, 9.4, 3.4, 1.3 and 7.4 μM for RNA1, RNA2, RNA3, RNA4 and RNA5, respectively.

Specifications

Chemical Name or Material MIR96-IN-1
CAS 1311982-88-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C33H48N8O2
Quantity 1 mg
Solubility Information DMSO : ≥ 100 mg/mL (169.84 mM)
SMILES O=C(NCCCN=[N+]=[N-])CCCOC1=C(C(C)(C)C)C=C(C2=NC3=CC=C(N4CCN(C)CC4)C=C3N2)C=C1C(C)(C)C
Molecular Weight (g/mol) 588.79
Formula Weight 588.79
Percent Purity 95.82%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.