missing translation for 'onlineSavingsMsg'
Learn More

Mirtazapine, MedChemExpress

Product Code. 16428256
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428256 100 mg 100mg
16402116 500 mg 500mg
16401416 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16428256 Supplier Medchem Express Supplier No. HYB0352
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Mirtazapine (Org3770) is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. Mirtazapine is also a 5-HT2, 5-HT3, histamine H1 receptor and α2-adrenoceptor antagonist with pKi values of 8.05, 8.1, 9.3 and 6.95, respectively.

Mirtazapine (Org3770) is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. Mirtazapine is also a 5-HT2, 5-HT3, histamine H1 receptor and α2-adrenoceptor antagonist with pKi values of 8.05, 8.1, 9.3 and 6.95, respectively.

Specifications

Chemical Name or Material Mirtazapine
CAS 85650-52-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H19N3
Quantity 100 mg
Synonym Org3770 6-Azamianserin
Solubility Information DMSO : 50 mg/mL (188.43 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.52 mM; Need ultrasonic)
SMILES CN1CC2C3=CC=CC=C3CC4=CC=CN=C4N2CC1
Molecular Weight (g/mol) 265.35
Formula Weight 265.35
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.