missing translation for 'onlineSavingsMsg'
Learn More

Mirtazapine N-Oxide, TRC

Product Code. 30135978
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135978 2.5 mg 2.5mg
17903085 10 mg 10mg
17932355 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135978 Supplier Toronto Research Chemicals Supplier No. TRCM3650102.5MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155172-12-6
Molecular Formula C17 H19 N3 O
Molecular Weight (g/mol) 281.35
Synonym (14bRS)-2-Methyl-1,2,3,4,10-14b-hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-Oxide, Mirtazapine Imp. A (EP), Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, 1,2,3,4,10,14b-hexahydro-2-methyl-, 2-oxide, 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-oxide
IUPAC Name (14bRS)-2-Methyl-1,2,3,4,10-14b-hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-oxide
SMILES C[N+]1([O-])CCN2C(C1)c3ccccc3Cc4cccnc24

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C17H19N3O/c1-20(21)10-9-19-16(12-20)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)19/h2-8,16H,9-12H2,1H3
Formula Weight 281.1528
Percent Purity >95%
Chemical Name or Material Mirtazapine N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.