missing translation for 'onlineSavingsMsg'
Learn More

Mirtazapine, TRC

Product Code. 17930785
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17930785 25 mg 25mg
30133814 100 mg 100mg
17922785 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17930785 Supplier Toronto Research Chemicals Supplier No. TRCM36500025MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85650-52-8
Molecular Formula C17 H19 N3
Molecular Weight (g/mol) 265.35
Synonym 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, (+/-)-1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, 6-Azamianserin, Mepirzapin, Mepirzepine, Mirtazapine, Mirtazepine, Mirtazipine, Org 3770, Promyrtil, Remergil, Remergon, Remeron, Rexer, Zispin
IUPAC Name 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine
SMILES CN1CCN2C(C1)c3ccccc3Cc4cccnc24

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3
Formula Weight 265.1579
Percent Purity >95%
Chemical Name or Material Mirtazapine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.