missing translation for 'onlineSavingsMsg'
Learn More

Misoprostol, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18762664
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18762664 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18762664 Supplier Tocris Bioscience Supplier No. 2297/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cytoprotective PGE1 analog

Cytoprotective prostaglandin E1 analog that displays agonist activity at EP receptors. Ki values are 120, 250, 67 and 67 nM at cloned mouse EP1, EP2, EP3 and EP4 receptors respectively. Prevents NSAID-induced gastric ulceration.
TRUSTED_SUSTAINABILITY

Specifications

CAS 59122-46-2
Molecular Formula C22H38O5
Quantity 10 mg
Synonym misoprostol, cytotec, isprelor, misoprostolum, misoprostil, misoprost, misotol, misoprostolum inn-latin, misodel, misogon
InChI Key OJLOPKGSLYJEMD-URPKTTJQSA-N
SMILES CCCCC(C)(CC=CC1C(CC(=O)C1CCCCCCC(=O)OC)O)O
IUPAC Name methyl 7-[(1R,2R,3R)-3-hydroxy-2-[(E)-4-hydroxy-4-methyloct-1-enyl]-5-oxocyclopentyl]heptanoate
Molecular Weight (g/mol) 382.541
PubChem CID 5282381
Formula Weight 382.53
Percent Purity >99%
Chemical Name or Material Misoprostol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.