missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mithramycin A
Description
Mithramycin A is an anticancer antibiotic that selectively binds to G-C-rich DNA in the presence of Mg2+ or Zn2+, inhibiting RNA and DNA polymerase action. Inhibits c-myc expression and induces myeloid differentiation of HL-60 promyelocytic leukemia cells.
Specifications
Specifications
| CAS | 18378-89-7 |
| Quantity | 1 mg |
| Solubility | Soluble to 50mM in DMSO |
| Product Line | Tocris |
| Molecular Formula | C52H76O24 |
| Formula Weight | 1085.16 |
| Product Type | Mithramycin A |
| Purity | >95% |
| Molecular Weight (g/mol) | 1085.156 |
| Chemical Name or Material | Mithramycin A |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?