missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Mitochondrial Membrane Potential Apoptosis Kit, with Mitotracker™ Red & Annexin V Alexa Fluor™ 488, for flow cytometry

Product Code. 10494012
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10494012 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10494012 Supplier Molecular Probes™ Supplier No. V35116
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mitochondrial Membrane Potential Apoptosis Kit, with Mitotracker™ Red & Annexin V Alexa Fluor™ 488, for flow cytometry

This flow cytometry product provides a rapid and convenient assay for two hallmarks of apoptosis - phosphatidylserine externalization and changes in mitochondrial membrane potential. After staining a cell population with Alexa Fluor™ 488 annexin V and MitoTracker™ Red CMXRos dye in the provided binding buffer, live cells exhibit very little green fluorescence and bright red fluorescence, whereas apoptotic cells exhibit green fluorescence and decreased red fluorescence. These populations can easily be distinguished using a flow cytometer, and the 488 nm line of an argon-ion laser can be used to excite both dyes.

View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Format Tube, Slide
Product Type Mitochondrial Membrane Apoptosis Kit
Content And Storage Contains 1 vial of annexin, Alexa Fluor™ 488 conjugate (250 μL), 3 vials of MitoTracker™ red (50 μg per vial), 1 bottle of annexin binding buffer (5X solution, 15 mL), and 1 vial of DMSO (100 μL).

Store in refrigerator (2–8°C) and protect from light.
Flow Cytometer Laser Lines 488
Excitation/Emission Mitotracker™ red 579⁄599, Alexa Fluor™ 488: 499⁄521
No. of Reactions 50
Shipping Condition Wet Ice
Product Line Alexa Fluor, MitoTracker
For Use With (Application) Flow Cytometry
Conjugate Alexa Fluor 488, MitoTracker Red
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.