missing translation for 'onlineSavingsMsg'
Learn More

Mitomycin A, TRC

Product Code. 30009060
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009060 25 mg 25mg
30004582 10 mg 10mg
30007923 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009060 Supplier Toronto Research Chemicals Supplier No. TRCM37189025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4055-39-4
Molecular Formula C16 H19 N3 O6
Molecular Weight (g/mol) 349.34
Synonym Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methyl-, (1aS,8S,8aR,8bS)-, Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 1,1a,2,8,8a,8b-hexahydro-8-(hydroxymethyl)-6,8a-dimethoxy-5-methyl-, carbamate (ester) (8CI), Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methyl-, [1aS-(1aα,8β,8aα,8bα)]-, Mitomycin A (7CI), (1aS,8S,8aR,8bS)-8-[[(Aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-6,8a-dimethoxy-5-methylazirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, NSC 75986, Mitomycin Imp. B (EP)
SMILES COC1=C(C)C(=O)C2=C([C@@H](COC(=O)N)[C@@]3(OC)[C@H]4N[C@H]4CN23)C1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H19N3O6/c1-6-11(20)10-9(12(21)13(6)23-2)7(5-25-15(17)22)16(24-3)14-8(18-14)4-19(10)16/h7-8,14,18H,4-5H2,1-3H3,(H2,17,22)/t7-,8+,14+,16-/m1/s1
Formula Weight 349.1274
Percent Purity >95%
Chemical Name or Material Mitomycin A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.